missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Individual Pipe Markers On A Card with Die-cut Arrowheads, with Pictograms - Gas

Product Code. 15399085 Shop All Brady Products
Change view
Click to view available options
Display Text:
Argon
Bio gas
CO2
Carbon dioxide
Condensate
Cooling gas
Exhaust
Freon
Freon 11
Freon 22
Gas
Helium
Inert gas
LPG
Mixed gas
Natural gas
Nitrogen
Recycled gas
Width (Metric):
250 mm
450 mm
Unit Size:
Pack of 4
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Display Text Width (Metric) Unit Size
15399085 Gas 250 mm -
15369025 Argon 250 mm -
15349045 Bio gas 250 mm -
15339095 Carbon dioxide 250 mm -
15319055 CO2 250 mm -
15339105 Cooling gas 250 mm -
15389055 Condensate 250 mm Pack of 4
15359065 Exhaust 250 mm Pack of 4
15379065 Exhaust 450 mm -
15399075 Freon 11 250 mm -
15339085 Freon 22 450 mm -
15319085 Freon 22 250 mm -
15339075 Freon 250 mm Pack of 4
15389125 Helium 250 mm -
15379105 Inert gas 250 mm Pack of 4
15339115 Natural gas 250 mm Pack of 4
15359125 LPG 250 mm -
15309115 Mixed gas 250 mm -
15369035 Nitrogen 250 mm -
15309125 Recycled gas 250 mm Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 options
This item is not returnable. View return policy
Product Code. 15399085 Supplier Brady™ Supplier No. N007512
  • ONLINE EXCLUSIVE PRICE
    17.90 €
    17.08 € / Pack of 4
    Save 0.82 €
    4% Off
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

European Standard Pipemarkers reinforce safety and aid maintenance.

  • Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
  • Danger and hazard symbols, if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, this saves you having to identify the appropriate symbol.
  • Markers are printed with a contrasting border in the same color as the text for improved visibility.
  • Individual markers are supplied with a die-cut arrow head on both ends: simply remove the one not required.

Specifications

Certifications/Compliance ROHS Environmental Compliance, EU Directive 2002/95/EC, European Directives 67/548 and 88/379
Description Individual Pipe Markers on a Card with die-cut arrowheads, with pictograms - Gas - Gas
Adhesive Type Permanent Acrylic, Permanent, Pressure-sensitive
Color Black on Ochre
Display Warning
Finish Gloss
Format Card
Material Polyester
Product Type Pipe Marker
Display Text Gas
For Use With (Application) Pipe and valve marking
Labels per Sheet 4
Languages English
No. per Pack 4 Markers/Card
Quantity 4 Pk.
Temperature Range (English) -40°F to +230°F
Temperature Range (Metric) -40°C to +110°C
Thickness (English) 0.0039 in.
Thickness (Metric) 0.099 mm
Dimensions (L x W) 26 x 250 mm
Length (English) 1.02 in.
Length (Metric) 26 mm
Width (English) 9.84 in.
Width (Metric) 250 mm
Height (Metric) Character 13 mm
Alternate Part Number GAS 250X26CARD-T1-P15
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.