missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Individual Pipe Markers On A Card with Die-cut Arrowheads - Steam

Product Code. 15359485 Shop All Brady Products
Change view
Click to view available options
Display Text:
HP Steam
High Pressure Steam
Low Pressure Steam
Overheated Steam
Saturated Steam
Steam
Width (Metric):
250 mm
450 mm
Unit Size:
Pack of 2
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Display Text Width (Metric) Unit Size
15359485 Steam 450 mm -
15359495 High Pressure Steam 250 mm -
15379495 High Pressure Steam 450 mm Pack of 2
15319505 HP Steam 250 mm -
15339505 HP Steam 450 mm Pack of 2
15319495 Low Pressure Steam 450 mm Pack of 2
15399485 Low Pressure Steam 250 mm -
15379505 Overheated Steam 250 mm -
15399505 Overheated Steam 450 mm -
15369505 Saturated Steam 450 mm Pack of 2
15349505 Saturated Steam 250 mm -
15339485 Steam 250 mm -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Product Code. 15359485 Supplier Brady™ Supplier No. N009449
  • ONLINE EXCLUSIVE PRICE
    14.45 €
    13.77 € / Pack of 2
    Save 0.68 €
    4% Off
Estimated Shipment: 17-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

European Standard Pipemarkers reinforce safety and aid maintenance.

  • Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
  • Danger and hazard symbols, if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, this saves you having to identify the appropriate symbol.
  • Markers are printed with a contrasting border in the same color as the text for improved visibility.
  • Individual markers are supplied with a die-cut arrow head on both ends: simply remove the one not required.

Specifications

Certifications/Compliance ROHS Environmental Compliance, EU Directive 2002/95/EC, European Directives 67/548 and 88/379
Description Individual Pipe Markers on a Card with die-cut arrowheads, without pictograms - Steam - Steam
Adhesive Type Permanent Acrylic, Permanent, Pressure-sensitive
Color Black on Silver
Finish Gloss
Format Card
Material Polyester
Product Type Pipe Marker
Display Text Steam
For Use With (Application) Pipe and valve marking
Labels per Sheet 2
Languages English
No. per Pack 2 Markers/Card
Quantity 2 Pk.
Temperature Range (English) -40°F to +230°F
Temperature Range (Metric) -40°C to +110°C
Thickness (English) 0.0039 in.
Thickness (Metric) 0.099 mm
Dimensions (L x W) 52 x 450 mm
Length (English) 2.05 in.
Length (Metric) 52 mm
Width (English) 17.72 in.
Width (Metric) 450 mm
Height (Metric) Character 22.5 mm
Alternate Part Number STEAM 450X52CARD-T1-P14
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.