missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Individual Pipe Markers On A Card with Die-cut Arrowheads, without Pictograms -- Air

Product Code. 15369235 Shop All Brady Products
Change view
Click to view available options
Display Text:
Air
Breathing air
Compressed air
Hot air
Instrument air
Return
Sterile air
Sterile compressed air
Vacuum
Unit Size:
Pack of 3
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Display Text Unit Size
15369235 Sterile air Pack of 3
15339205 Air Pack of 3
15359215 Compressed air Pack of 3
15309235 Breathing air Pack of 3
15399205 Hot air -
15349225 Instrument air -
15399235 Return Pack of 3
15319225 Sterile compressed air Pack of 3
15329245 Vacuum -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 15369235 Supplier Brady™ Supplier No. N008467
  • ONLINE EXCLUSIVE PRICE
    16.95 €
    16.31 € / Pack of 3
    Save 0.64 €
    3% Off
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

European Standard Pipemarkers reinforce safety and aid maintenance.

  • Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
  • Danger and hazard symbols, if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, this saves you having to identify the appropriate symbol.
  • Markers are printed with a contrasting border in the same color as the text for improved visibility.
  • Individual markers are supplied with a die-cut arrow head on both ends: simply remove the one not required.

Specifications

Certifications/Compliance ROHS Environmental Compliance, EU Directive 2002/95/EC, European Directives 67/548 and 88/379
Description Individual Pipe Markers on a Card with die-cut arrowheads, without pictograms - Air - Sterile air
Adhesive Type Permanent Acrylic, Permanent, Pressure-sensitive
Color White on Blue
Finish Gloss
Format Card
Material Polyester
Product Type Pipe Marker
Display Text Sterile air
For Use With (Application) Pipe and valve marking
Labels per Sheet 3
Languages English
No. per Pack 3 Markers/Card
Quantity 3 Pk.
Temperature Range (English) -40°F to +230°F
Temperature Range (Metric) -40°C to +110°C
Thickness (English) 0.0039 in.
Thickness (Metric) 0.099 mm
Dimensions (L x W) 37 x 335 mm
Width (English) 13.98 in.
Width (Metric) 355 mm
Height (English) 1.46 in.
Height (Metric) 37 mm
Height (Metric) Character 16.5 mm
Alternate Part Number STERILE AIR 355X37CARD-T1-P8
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.