missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Individual Pipe Markers On A Card with Die-cut Arrowheads, without Pictograms - Fire Fighting

Product Code. 15359245
Change view
Click to view available options
Display Text:
Fire protection Water
Foam
Sprinkler
Sprinkler Water
Width (Metric):
250 mm
450 mm
Unit Size:
Pack of 2
Pack of 4
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Display Text Width (Metric) unitSize
15359245 Fire protection Water 250 mm -
15379245 Fire protection Water 450 mm Pack of 2
15389255 Foam 250 mm -
15309265 Foam 450 mm Pack of 2
15329255 Sprinkler Water 250 mm Pack of 4
15349255 Sprinkler Water 450 mm Pack of 2
15349265 Sprinkler 250 mm -
15369265 Sprinkler 450 mm Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 15359245 Supplier Brady™ Supplier No. N008529
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

European Standard Pipemarkers reinforce safety and aid maintenance.

  • Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
  • Danger and hazard symbols, if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, this saves you having to identify the appropriate symbol.
  • Markers are printed with a contrasting border in the same color as the text for improved visibility.
  • Individual markers are supplied with a die-cut arrow head on both ends: simply remove the one not required.

Specifications

Certifications/Compliance ROHS Environmental Compliance, EU Directive 2002/95/EC, European Directives 67/548 and 88/379
Description Individual Pipe Markers on a Card with die-cut arrowheads, without pictograms - Fire Fighting - Fire protection water
Adhesive Type Permanent Acrylic, Permanent, Pressure-sensitive
Color White on Red
Finish Gloss
Format Card
Material Polyester
Product Type Pipe Marker
Display Text Fire protection Water
For Use With (Application) Pipe and valve marking
Labels per Sheet 4
Languages English
No. per Pack 4 Markers/Card
Quantity 4 Pk.
Temperature Range (English) -40°F to +230°F
Temperature Range (Metric) -40°C to +110°C
Thickness (English) 0.0039 in.
Thickness (Metric) 0.099 mm
Dimensions (L x W) 26 x 250 mm
Length (English) 1.02 in.
Length (Metric) 26 mm
Width (English) 9.84 in.
Width (Metric) 250 mm
Height (Metric) Character 13.3 mm
Alternate Part Number FIRE PROTECTION WATER 250X26CARD-T1-P04
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.