missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Individual Pipe Markers On A Card with Die-cut Arrowheads, without Pictograms - Flammable/Non Flammable Liquids/Oils

Product Code. 15389135
Change view
Click to view available options
Display Text:
Emulsion
Hydraulic oil
Lubricating oil
Oil
Radioactive Return
Residue
Used solvents
Width (Metric):
250 mm
450 mm
Unit Size:
Pack of 2
Pack of 4
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Display Text Width (Metric) unitSize
15389135 Emulsion 450 mm Pack of 2
15369135 Emulsion 250 mm Pack of 4
15309185 Hydraulic oil 250 mm -
15329185 Hydraulic oil 450 mm Pack of 2
15349175 Lubricating oil 250 mm -
15369175 Lubricating oil 450 mm -
15389165 Oil 250 mm -
15309175 Oil 450 mm -
15399185 Radioactive Return 250 mm -
15329195 Radioactive Return 450 mm Pack of 2
15369185 Residue 250 mm -
15389185 Residue 450 mm -
15339195 Used solvents 250 mm Pack of 4
15359195 Used solvents 450 mm Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
This item is not returnable. View return policy
Product Code. 15389135 Supplier Brady™ Supplier No. N007920
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

European Standard Pipemarkers reinforce safety and aid maintenance.

  • Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
  • Danger and hazard symbols, if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, this saves you having to identify the appropriate symbol.
  • Markers are printed with a contrasting border in the same color as the text for improved visibility.
  • Individual markers are supplied with a die-cut arrow head on both ends: simply remove the one not required.

Specifications

Certifications/Compliance ROHS Environmental Compliance, EU Directive 2002/95/EC, European Directives 67/548 and 88/379
Description Individual Pipe Markers on a Card with die-cut arrowheads, without pictograms - Flammable/Non Flammable Liquids/Oils - Emulsion
Adhesive Type Permanent Acrylic, Permanent, Pressure-sensitive
Color White on Dark Brown
Finish Gloss
Format Card
Material Polyester
Product Type Pipe Marker
Display Text Emulsion
For Use With (Application) Pipe and valve marking
Labels per Sheet 2
Languages English
No. per Pack 2 Markers/Card
Quantity 2 Pk.
Temperature Range (English) -40°F to +230°F
Temperature Range (Metric) -40°C to +110°C
Thickness (English) 0.0039 in.
Thickness (Metric) 0.099 mm
Dimensions (L x W) 52 x 450 mm
Length (English) 2.05 in.
Length (Metric) 52 mm
Width (English) 17.72 in.
Width (Metric) 450 mm
Height (Metric) Character 22.5 mm
Alternate Part Number EMULSION 450X52CARD-T1-P19
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.