missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Individual Pipe Markers On A Card with Die-cut Arrowheads, without Pictograms - Water

Product Code. 15399465 Shop All Brady Products
Change view
Click to view available options
Display Text:
Boiler feed water
Brine Water
Canal Water
Chilled Water
Chlorated Water
City Water
Cold Water Return
Cold Water supply
Condensate
Condenser Water
Condenser Water Return
Condenser Water supply
Cooling Water
Decarbonatized Water
Deionized Water
Demineralised Water
Distilled Water
Drinking Water
Filtered Water
Heating
Heating Return
Heating Water
Heating supply
Hot Water
Hot water return
Industrial Water
Low pressure Water
Mineral Water
Non-drinking Water
Plant Water
Polluted Water
Process Water
Pure Water
Purified Water
Raw Water
Return
River Water
Secondary circuit
Soft Water
Supply
Treated Water
Waste Water
Water
Unit Size:
Pack of 3
43 product options available for selection
Product selection table with 43 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Display Text Unit Size
15399465 Waste Water -
15389305 Boiler feed water Pack of 3
15309465 Brine Water -
15329405 Canal Water Pack of 3
15349455 Chilled Water Pack of 3
15369335 Chlorated Water -
15309375 City Water -
15349415 Cold Water Return -
15339295 Condensate Pack of 3
15369345 Condenser Water -
15389405 Cold Water supply Pack of 3
15329345 Condenser Water Return Pack of 3
15399345 Condenser Water supply Pack of 3
15319365 Cooling Water -
15339375 Decarbonatized Water -
15369375 Deionized Water Pack of 3
15369385 Distilled Water Pack of 3
15399435 Drinking Water Pack of 3
15309385 Demineralised Water Pack of 3
15399395 Filtered Water Pack of 3
15349275 Heating -
15339285 Heating Return -
15379275 Heating supply -
15329355 Heating Water -
15319325 Hot Water -
15309335 Hot water return Pack of 3
15319425 Industrial Water -
15319315 Low pressure Water -
15309435 Non-drinking Water -
15379425 Mineral Water -
15359475 Plant Water Pack of 3
15369435 Polluted Water Pack of 3
15359445 Process Water Pack of 3
15369395 Pure Water -
15319455 Purified Water -
15359315 Raw Water Pack of 3
15389475 Return -
15399385 Soft Water -
15379365 River Water -
15309295 Secondary circuit Pack of 3
15389265 Supply -
15339465 Treated Water Pack of 3
15319305 Water -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 43 options available.
43 options
This item is not returnable. View return policy
Product Code. 15399465 Supplier Brady™ Supplier No. N009394
  • ONLINE EXCLUSIVE PRICE
    17.05 €
    16.31 € / Pack of 3
    Save 0.74 €
    4% Off
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

European Standard Pipemarkers reinforce safety and aid maintenance.

  • Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
  • Danger and hazard symbols, if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, this saves you having to identify the appropriate symbol.
  • Markers are printed with a contrasting border in the same color as the text for improved visibility.
  • Individual markers are supplied with a die-cut arrow head on both ends: simply remove the one not required.

Specifications

Certifications/Compliance ROHS Environmental Compliance, EU Directive 2002/95/EC, European Directives 67/548 and 88/379
Description Individual Pipe Markers on a Card with die-cut arrowheads, without pictograms - Water - Waste water
Adhesive Type Permanent Acrylic, Permanent, Pressure-sensitive
Color White on Green
Finish Gloss
Format Card
Material Polyester
Product Type Pipe Marker
Display Text Waste Water
For Use With (Application) Pipe and valve marking
Labels per Sheet 3
Languages English
No. per Pack 3 Markers/Card
Quantity 3 Pk.
Temperature Range (English) -40°F to +230°F
Temperature Range (Metric) -40°C to +110°C
Thickness (English) 0.0039 in.
Thickness (Metric) 0.099 mm
Dimensions (L x W) 37 x 335 mm
Width (English) 13.98 in.
Width (Metric) 355 mm
Height (English) 1.46 in.
Height (Metric) 37 mm
Height (Metric) Character 16.5 mm
Alternate Part Number WASTE WATER 355X37CARD-T1-P03
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.