missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Individual Pipe Markers On A Card with Die-cut Arrowheads, without Pictograms - Water (A-H)

Product Code. 15359275 Shop All Brady Products
Change view
Click to view available options
Display Text:
Boiler feed water
Brine water
Canal water
Chilled water
Chlorated water
City water
Cold water return
Cold water supply
Condensate
Condenser water
Condenser water return
Condenser water supply
Cooling water
Decarbonatized Water
Deionized water
Demineralised Water
Distilled water
Drinking water
Filtered water
Heating
Heating return
Heating supply
Heating water
Hot water
Hot water return
Width (Metric):
250 mm
450 mm
Unit Size:
Pack of 2
Pack of 4
50 product options available for selection
Product selection table with 50 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Display Text Width (Metric) Unit Size
15359275 Heating 450 mm Pack of 2
15379305 Boiler feed water 250 mm Pack of 4
15399305 Boiler feed water 450 mm -
15399455 Brine water 250 mm Pack of 4
15319465 Brine water 450 mm Pack of 2
15319405 Canal water 250 mm -
15339405 Canal water 450 mm Pack of 2
15339455 Chilled water 250 mm -
15359455 Chilled water 450 mm -
15359335 Chlorated water 250 mm Pack of 4
15319375 City water 450 mm Pack of 2
15399365 City water 250 mm -
15379335 Chlorated water 450 mm Pack of 2
15339415 Cold water return 250 mm -
15359415 Cold water return 450 mm Pack of 2
15399405 Cold water supply 450 mm -
15379405 Cold water supply 250 mm Pack of 4
15329295 Condensate 250 mm Pack of 4
15349295 Condensate 450 mm -
15319345 Condenser water return 250 mm Pack of 4
15339345 Condenser water return 450 mm -
15389345 Condenser water supply 250 mm Pack of 4
15309355 Condenser water supply 450 mm -
15349345 Condenser water 250 mm -
15379345 Condenser water 450 mm -
15309365 Cooling water 250 mm -
15329365 Cooling water 450 mm Pack of 2
15329375 Decarbonatized Water 250 mm -
15349375 Decarbonatized Water 450 mm -
15359375 Deionized water 250 mm Pack of 4
15389375 Demineralised Water 250 mm Pack of 4
15379375 Deionized water 450 mm Pack of 2
15319385 Demineralised Water 450 mm Pack of 2
15359385 Distilled water 250 mm Pack of 4
15379385 Distilled water 450 mm -
15389435 Drinking water 250 mm Pack of 4
15309445 Drinking water 450 mm Pack of 2
15309405 Filtered water 450 mm Pack of 2
15389395 Filtered water 250 mm Pack of 4
15329285 Heating return 250 mm Pack of 4
15369275 Heating supply 250 mm Pack of 4
15349285 Heating return 450 mm Pack of 2
15319355 Heating water 250 mm -
15389275 Heating supply 450 mm Pack of 2
15339355 Heating water 450 mm Pack of 2
15339275 Heating 250 mm -
15319335 Hot water return 450 mm -
15399325 Hot water return 250 mm Pack of 4
15309325 Hot water 250 mm -
15329325 Hot water 450 mm -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 50 options available.
50 options
This item is not returnable. View return policy
Product Code. 15359275 Supplier Brady™ Supplier No. N008603
  • ONLINE EXCLUSIVE PRICE
    14.45 €
    13.77 € / Pack of 2
    Save 0.68 €
    4% Off
Estimated Shipment: 17-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

European Standard Pipemarkers reinforce safety and aid maintenance.

  • Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
  • Danger and hazard symbols, if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, this saves you having to identify the appropriate symbol.
  • Markers are printed with a contrasting border in the same color as the text for improved visibility.
  • Individual markers are supplied with a die-cut arrow head on both ends: simply remove the one not required.

Specifications

Certifications/Compliance ROHS Environmental Compliance, EU Directive 2002/95/EC, European Directives 67/548 and 88/379
Description Individual Pipe Markers on a Card with die-cut arrowheads, without pictograms - Water - Heating
Adhesive Type Permanent Acrylic, Permanent, Pressure-sensitive
Color White on Green
Finish Gloss
Format Card
Material Polyester
Product Type Pipe Marker
Display Text Heating
For Use With (Application) Pipe and valve marking
Labels per Sheet 2
Languages English
No. per Pack 2 Markers/Card
Quantity 2 Pk.
Temperature Range (English) -40°F to +230°F
Temperature Range (Metric) -40°C to +110°C
Thickness (English) 0.0039 in.
Thickness (Metric) 0.099 mm
Dimensions (L x W) 52 x 450 mm
Length (English) 2.05 in.
Length (Metric) 52 mm
Width (English) 17.72 in.
Width (Metric) 450 mm
Height (Metric) Character 22.5 mm
Alternate Part Number HEATING 450X52CARD-T1-P03
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.