missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Individual Pipe Markers On A Card with Die-cut Arrowheads, without Pictograms - Water (I-Z)

Product Code. 15399475 Shop All Brady Products
Change view
Click to view available options
Display Text:
Industrial water
Low pressure water
Mineral water
Non-drinking water
Plant water
Polluted water
Primary circuit
Process water
Pure water
Purified water
Raw water
Return
River water
Secondary circuit
Soft water
Supply
Treated water
Waste water
Water
Well water
Width (Metric):
250 mm
450 mm
Unit Size:
Pack of 2
Pack of 4
39 product options available for selection
Product selection table with 39 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Display Text Width (Metric) Unit Size
15399475 Return 450 mm -
15309425 Industrial water 250 mm -
15329425 Industrial water 450 mm -
15309315 Low pressure water 250 mm Pack of 4
15339315 Low pressure water 450 mm Pack of 2
15369425 Mineral water 250 mm -
15389425 Mineral water 450 mm -
15319435 Non-drinking water 450 mm -
15399425 Non-drinking water 250 mm Pack of 4
15349475 Plant water 250 mm -
15369475 Plant water 450 mm Pack of 2
15359435 Polluted water 250 mm Pack of 4
15379435 Polluted water 450 mm Pack of 2
15389285 Primary circuit 250 mm Pack of 4
15349445 Process water 250 mm Pack of 4
15369445 Process water 450 mm -
15359395 Pure water 250 mm Pack of 4
15379395 Pure water 450 mm Pack of 2
15309455 Purified water 250 mm -
15349315 Raw water 250 mm Pack of 4
15369315 Raw water 450 mm Pack of 2
15329455 Purified water 450 mm Pack of 2
15379475 Return 250 mm Pack of 4
15369365 River water 250 mm -
15389365 River water 450 mm -
15399285 Secondary circuit 250 mm -
15319295 Secondary circuit 450 mm Pack of 2
15389385 Soft water 250 mm Pack of 4
15379265 Supply 250 mm -
15309395 Soft water 450 mm -
15399265 Supply 450 mm Pack of 2
15329465 Treated water 250 mm -
15349465 Treated water 450 mm -
15389465 Waste water 250 mm Pack of 4
15309475 Waste water 450 mm Pack of 2
15369355 Well water 450 mm Pack of 2
15349355 Well water 250 mm -
15399295 Water 250 mm -
15329305 Water 450 mm -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 39 options available.
39 options
This item is not returnable. View return policy
Product Code. 15399475 Supplier Brady™ Supplier No. N009422
  • ONLINE EXCLUSIVE PRICE
    14.45 €
    13.77 € / Pack of 2
    Save 0.68 €
    4% Off
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

European Standard Pipemarkers reinforce safety and aid maintenance.

  • Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
  • Danger and hazard symbols, if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, this saves you having to identify the appropriate symbol.
  • Markers are printed with a contrasting border in the same color as the text for improved visibility.
  • Individual markers are supplied with a die-cut arrow head on both ends: simply remove the one not required.

Specifications

Certifications/Compliance ROHS Environmental Compliance, EU Directive 2002/95/EC, European Directives 67/548 and 88/379
Description Individual Pipe Markers on a Card with die-cut arrowheads, without pictograms - Water - Return
Adhesive Type Permanent Acrylic, Permanent, Pressure-sensitive
Color White on Green
Finish Gloss
Format Card
Material Polyester
Product Type Pipe Marker
Display Text Return
For Use With (Application) Pipe and valve marking
Labels per Sheet 2
Languages English
No. per Pack 2 Markers/Card
Quantity 2 Pk.
Temperature Range (English) -40°F to +230°F
Temperature Range (Metric) -40°C to +110°C
Thickness (English) 0.0039 in.
Thickness (Metric) 0.099 mm
Dimensions (L x W) 52 x 450 mm
Length (English) 2.05 in.
Length (Metric) 52 mm
Width (English) 17.72 in.
Width (Metric) 450 mm
Height (Metric) Character 22.5 mm
Alternate Part Number RETURN 450X52CARD-T1-P03
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.