Learn More
Fisherbrand™ Individual Stainless Steel Cylindrical Calibration Weights, Class M1 with certificate
Shop All Fisherbrand Traceable ProductsSpecifications
Specifications
| Classification | M1 |
| Material | Stainless Steel |
| Weight (Metric) | 10 g |
| Mass | 10 g |
| Shape | Cylindrical |