Learn More
Fisherbrand™ Individual Stainless Steel Leaf Calibration Weights, Class E2
Product Code. 15535089 Shop All Fisherbrand Products
Change view
Click to view available options
Weight (Metric):
1 mg
2 mg
5 mg
10 mg
20 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
1 unit
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Weight (Metric) unitSize
15535089 2 mg 1 unit
15575089 50 mg 1 unit
15585089 100 mg 1 unit
15595089 200 mg 1 unit
15545089 5 mg 1 unit
15555089 10 mg 1 unit
15565089 20 mg 1 unit
15525089 1 mg 1 unit
15505099 500 mg 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 15535089
  • ONLINE EXCLUSIVE PRICE
    38.20 €
    26.15 € / 1 units
    Save 12.05 €
    31% Off
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
Calibrate weights of lower accuracy classes. Fisherbrand™ Class E2 Individual Stainless Steel Leaf Calibration Weights are intended for use in the verification or calibration of class F1 weights and for use with weighing instruments of special accuracy class I.
  • Class E2 calibration weight
  • In compliance with OIML R111
Specifications
Classification E2
Material Stainless Steel
Weight (Metric) 2 mg
Mass 2 mg