missing translation for 'onlineSavingsMsg'
Learn More

Indole-2-carboxylic acid, 99%

Product Code. 10785001 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
Product Code. Quantity Packaging unitSize
10785001 50 g Glass bottle 50g
10676312 10 g Glass bottle 10g
2 options
This item is not returnable. View return policy

Product Code. 10785001

Brand: Thermo Scientific Acros 122200500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1477-50-5 | C9H7NO2 | 161.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1477-50-5
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD00005611
InChI Key HCUARRIEZVDMPT-UHFFFAOYSA-N
Synonym indole-2-carboxylic acid, 2-carboxyindole, 2-indolecarboxylic acid, indol-2-carboxylic acid, 1h-indolecarboxylic acid, indole-2-carboxylate, 2-indolylformic acid, 2-indole carboxylic acid, 1h-indol-2-carbons, carboxyindole
PubChem CID 72899
IUPAC Name 1H-indole-2-carboxylic acid
SMILES OC(=O)C1=CC2=CC=CC=C2N1

Specifications

Melting Point 205.0°C to 209.0°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 50 g
Beilstein 22, 61
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,ether
Formula Weight 161.16
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Indole-2-carboxylic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Indole-2-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.