missing translation for 'onlineSavingsMsg'
Learn More

Indole-3-Butyric Acid , MP Biomedicals, LLC

Product Code. 11328879 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 g
Packaging:
Glass
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11328879 50 g Glass 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11328879 Supplier MP Biomedicals Supplier No. 0210204250
 Restricted Item
Estimated Shipment: 01-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Indole-3-butyric has a purity of 99.2% acid is used as plant growth hormone.

  • A naturally occurring phytohormone auxin (plant growth regulator)
  • Promotes root formation in cuttings, but does not affect ethylene levels.

Specifications

CAS 133-32-4
Melting Point 118°C to 126°C
Color White
Packaging Glass
Molecular Formula C12H13NO2
Quantity 50 g
Synonym indole-3-butyric acid, 3-indolebutyric acid, 4-1h-indol-3-yl butanoic acid, indolebutyric acid, hormodin, 1h-indole-3-butanoic acid, seradix, indole-3-butanoic acid, jiffy grow, 4-indol-3-yl butyric acid
InChI Key JTEDVYBZBROSJT-UHFFFAOYSA-N
Molecular Weight (g/mol) 203.24
PubChem CID 8617
ChEBI CHEBI:33070
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Indole-3-butyric Acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 4-(indol-3-yl)butyric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.