missing translation for 'onlineSavingsMsg'
Learn More

Indole-3-carboxylic acid, MedChemExpress

Product Code. 16497816
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16497816 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16497816 Supplier Medchem Express Supplier No. HY40161
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Indole-3-carboxylic acid is a normal urinary indolic tryptophan metabolite and has been found elevated in patients with liver diseases.

Indole-3-carboxylic acid is a normal urinary indolic tryptophan metabolite and has been found elevated in patients with liver diseases.

Specifications

Chemical Name or Material Indole-3-carboxylic acid
CAS 771-50-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C9H7NO2
Solubility Information DMSO : 100 mg/mL (620.50 mM; Need ultrasonic)
SMILES O=C(O)C1=CNC2=CC=CC=C12
Molecular Weight (g/mol) 161.16
Quantity 100 mg
Formula Weight 161.16
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.