missing translation for 'onlineSavingsMsg'
Learn More

Indole-3-propionic acid, 98%

Product Code. 11371737
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11371737 25 g 25g
11361737 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11371737 Supplier Thermo Scientific Alfa Aesar Supplier No. L04877.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 830-96-6 | C11H11NO2 | 189.21 g/mol

Indole-3-propionic acid is a reactant for preparation of fluorescent analogues of strigolactones, melanocortin receptors ligands, histamine H4 receptor agonists, and NR2B/NMDA receptor antagonists. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used to Study as an adjunct to improve perfusion after liver transplant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Indole-3-propionic acid is a reactant for preparation of fluorescent analogues of strigolactones, melanocortin receptors ligands, histamine H4 receptor agonists, and NR2B/NMDA receptor antagonists. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used to Study as an adjunct to improve perfusion after liver transplant.

Solubility
It is slightly soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 830-96-6
Molecular Formula C11H11NO2
Molecular Weight (g/mol) 189.21
MDL Number MFCD00005660
InChI Key GOLXRNDWAUTYKT-UHFFFAOYSA-N
Synonym 3-indolepropionic acid, indole-3-propionic acid, 3-1h-indol-3-yl propanoic acid, 1h-indole-3-propanoic acid, indolepropionic acid, indolylpropionic acid, 3-3-indolyl propionic acid, 3-3-indolyl propanoic acid, ipa auxin, 1h-indole-3-propionic acid
PubChem CID 3744
ChEBI CHEBI:43580
IUPAC Name 3-(1H-indol-3-yl)propanoic acid
SMILES OC(=O)CCC1=CNC2=CC=CC=C12

Specifications

Melting Point 132°C to 135°C
Quantity 25 g
Beilstein 147733
Solubility Information It is slightly soluble in water.
Formula Weight 189.21
Percent Purity 98%
Chemical Name or Material Indole-3-propionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.