missing translation for 'onlineSavingsMsg'
Learn More

Indole-4-methanol, 97%, Thermo Scientific™

Product Code. 15325018 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15325018 250 mg 250mg
15315018 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15325018 Supplier Thermo Scientific Acros Supplier No. 455532500
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 1074-85-7
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.18
MDL Number MFCD01632220
InChI Key BVSGXWCTWBZFEV-UHFFFAOYSA-N
Synonym indole-4-methanol, 1h-indol-4-yl methanol, 1h-indole-4-methanol, 4-hydroxymethylindole, 4-hydroxymethyl-1h-indole, indol-4-ylmethan-1-ol, pubchem7973, 4-hydroxymethyl indole, 4-hydroxymethyl-indole, acmc-2098wi
PubChem CID 2773457
IUPAC Name 1H-indol-4-ylmethanol
SMILES OCC1=C2C=CNC2=CC=C1

Specifications

Melting Point 46.0°C to 50.0°C
Assay Percent Range 96% min. (GC)
Quantity 250 mg
Formula Weight 147.17
Percent Purity 97%
Chemical Name or Material Indole-4-methanol
compliance-icons
Product Identifier
  • Indol-4-methanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.