missing translation for 'onlineSavingsMsg'
Learn More

Indole-7-carboxylic acid, 97%

Product Code. 15491068
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15491068 1 g 1g
15401078 5 g 5g
15411078 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15491068 Supplier Thermo Scientific Alfa Aesar Supplier No. H54931.03
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1670-83-3 | C9H7NO2 | 161.16 g/mol

Indole-7-carboxylic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Indole-7-carboxylic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1670-83-3
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD00210442
InChI Key IPDOBVFESNNYEE-UHFFFAOYSA-N
Synonym indole-7-carboxylic acid, 7-indolecarboxylic acid, indole-7-carboxylicacid, zlchem 444, pubchem2475, 7-carboxyindole, acmc-209dvl, indole-7-carboxlic acid, indole 7-carboxylic acid, 1h-indole-7-carboxylicacid
PubChem CID 74281
IUPAC Name 1H-indole-7-carboxylic acid
SMILES C1=CC2=C(C(=C1)C(=O)O)NC=C2

Specifications

Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 161.05
Percent Purity 97%
Chemical Name or Material Indole-7-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.