missing translation for 'onlineSavingsMsg'
Learn More

Indole, 99+%

Product Code. 10205310 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
10g
2.5kg
25g
500g
This item is not returnable. View return policy

Product Code. 10205310

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 120-72-9 | C8H7N | 117.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-72-9
MDL Number MFCD00005607
InChI Key SIKJAQJRHWYJAI-UHFFFAOYSA-N
Synonym indole, indol, 2,3-benzopyrrole, 1-benzazole, ketole, 1-azaindene, benzopyrrole, 2,3-benzopyrole, caswell no. 498b, indol german
PubChem CID 798
ChEBI CHEBI:16881
IUPAC Name 1H-indole
SMILES C1=CC=C2C(=C1)C=CN2

Specifications

Melting Point 51.0°C to 53.0°C
Color Pink to White
Density 1.2200g/mL
Boiling Point 253.0°C to 254.0°C
Flash Point 136°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass Bottle
Molecular Formula C8H7N
Quantity 100 g
Beilstein 20, 304
Merck Index 15, 5003
Specific Gravity 1.22
Solubility Information Solubility in water: 2.80g/L (25°C). Other solubilities: soluble in hot water,soluble in hot alcohol,ether and benzene,0.1g/mL meoh
Molecular Weight (g/mol) 117.15
Formula Weight 117.15
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Indole, 98+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Indole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Indole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.