missing translation for 'onlineSavingsMsg'
Learn More

Indoleacetic Acid, TRC

Product Code. 30134492
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134492 25 g 25g
17947649 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134492 Supplier Toronto Research Chemicals Supplier No. TRCI57734025G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-51-4
Molecular Formula C10 H9 N O2
Molecular Weight (g/mol) 175.18
Synonym 1H-Indole-3-acetic acid, Indole-3-acetic acid (8CI), (1H-Indol-3-yl)acetic acid, 2-(1H-Indol-3-yl)acetic acid, 2-(3-Indolyl)acetic acid, 3-(Carboxymethyl)-1H-indole, 3-(Carboxymethyl)indole, 3-IAA, 3-Indolylmethylcarboxylic acid, Bioenraiz, GAP, Heteroauxin, IAA, Indoleacetic acid, NSC 3787, Noclosan, Rhizopin, α-IAA, β-IAA, β-Indoleacetic acid, 3-Indolylacetic acid, 3-Indolyl acetic acid
IUPAC Name 2-(1H-indol-3-yl)acetic acid
SMILES OC(=O)Cc1c[nH]c2ccccc12

Specifications

Quantity 25 g
InChI Formula InChI=1S/C10H9NO2/c12-10(13)5-7-6-11-9-4-2-1-3-8(7)9/h1-4,6,11H,5H2,(H,12,13)
Formula Weight 175.0633
Chemical Name or Material 3-Indolyl-acetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.