missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Indomethacin

Product Code. 17190292
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17190292 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17190292 Supplier STEMCELL Technologies Supplier No. 73942
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Inhibits COX-1 and COX-2

  • Indomethacin is a non-steroidal, anti-inflammatory agent that inhibits cyclooxygenase (COX) activity, thereby blocking the production of prostaglandins (Vane et al.) Indomethacin inhibits both COX-1 and COX-2 (IC50 = 0.08 and 0.96 μM for recombinant human COX-1 and COX-2, respectively; Kurumbail et al.)
  • Activates peroxisome proliferator-activated receptor-γ (PPARγ), a ligand-activated transcription factor known to play a pivotal role in adipogenesis (Lehmann et al.)
  • Inhibits chondrogenic differentiation in ATDC5 cells and bone marrow stem cells (Caron et al.)
  • Inhibits growth of mouse mammary tumors (Fulton)
  • Induces apoptosis in prostate and gastric cancer cells (Chiou et al.; Liu et al.)

Specifications

CAS 53-86-1
Synonym Indocin
Quantity 1 g
Molecular Formula C19H16ClNO4
Purity ≥99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.