missing translation for 'onlineSavingsMsg'
Learn More

Indomethacin Isopropyl Ester, TRC

Product Code. 30395977
Change view
Click to view available options
Quantity:
500 mg
1 g
2.5 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30395977 1 g 1g
30391351 500 mg -
30405324 2.5 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30395977 Supplier Toronto Research Chemicals Supplier No. TRCI4621301G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72616-25-2
Molecular Formula C22 H22 Cl N O4
Molecular Weight (g/mol) 399.87
Synonym 1-(4-Chlorobenzoyl)-5-methoxy-2-methyl-1H-indole-3-acetic acid 1-methylethyl ester
IUPAC Name propan-2-yl 2-[1-(4-chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetate
SMILES COc1ccc2c(c1)c(CC(=O)OC(C)C)c(C)n2C(=O)c3ccc(Cl)cc3

Specifications

Quantity 1 g
InChI Formula InChI=1S/C22H22ClNO4/c1-13(2)28-21(25)12-18-14(3)24(20-10-9-17(27-4)11-19(18)20)22(26)15-5-7-16(23)8-6-15/h5-11,13H,12H2,1-4H3
Formula Weight 399.1237
Chemical Name or Material Indomethacin Isopropyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.