missing translation for 'onlineSavingsMsg'
Learn More

Indomethacin, MedChemExpress

Product Code. 16471409
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16471409 1 g 1g
16448657 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16471409 Supplier Medchem Express Supplier No. HY14397
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Indomethacin (Indometacin) is a potent, orally active COX1/2 inhibitor with IC50 values of 18 nM and 26 nM for COX-1 and COX-2, respectively. Indomethacin has anticancer activity and anti-infective activity. Indomethacin can be used for cancer, inflammation and viral infection research.

Indomethacin (Indometacin) is a potent, orally active COX1/2 inhibitor with IC50 values of 18 nM and 26 nM for COX-1 and COX-2, respectively. Indomethacin has anticancer activity and anti-infective activity. Indomethacin can be used for cancer, inflammation and viral infection research.

Specifications

Chemical Name or Material Indomethacin
CAS 53-86-1
Color White
Purity Grade Notes Research
Molecular Formula C19H16ClNO4
Quantity 1 g
Synonym Indometacin
Solubility Information DMSO : 100 mg/mL (279.49 mM; Need ultrasonic) ∣Ethanol : 12.5 mg/mL (34.94 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES COC1=CC=C(N(C(C2=CC=C(Cl)C=C2)=O)C(C)=C3CC(O)=O)C3=C1
Molecular Weight (g/mol) 357.79
Formula Weight 357.79
Percent Purity 99.71%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.