missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Indomethacin

Product Code. 18702754
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18702754 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18702754 Supplier R&D Systems™ Supplier No. 1708/100
  • ONLINE EXCLUSIVE PRICE
    171.89 €
    154.35 € / 100mg
    Save 17.54 €
    10% Off
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Cyclooxygenase inhibitor (COX-1 > COX-2)

Cyclooxgenase (COX) inhibitor; displays selectivity for COX-1 (IC50 values are 230 and 630 nM for human COX-1 and COX-2 respectively). Widely used anti-inflammatory agent. Identified by chemoinformatics as targeting human host proteins that interact with SARS-CoV-2.
TRUSTED_SUSTAINABILITY

Specifications

CAS 53-86-1
Quantity 100 mg
Product Line Tocris
Molecular Formula C19H16ClNO4
Formula Weight 357.79
Purity >99%
Molecular Weight (g/mol) 357.79
Chemical Name or Material Indomethacin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.