missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Indubiose A37

Product Code. 15541441 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15541441 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15541441 Supplier MP Biomedicals™ Supplier No. 11AGAI0025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Indubiose A37

TRUSTED_SUSTAINABILITY

Specifications

CAS 9012-36-6
Description Standard agarose is a purified lineargalactan hydrocolloid isolated from agar or agar-bearing marine algae with an Electroendoosmosis (EEO) value of 0.12 making it an essential component in chemical separation techniques.
For Use With (Application) Electrophoresis; Separation of DNA/RNA fragments from 400 to 8000 bp
Melting Temperature 86.5-89.5°C (1.5%gel)
Quantity 25 g
Storage Requirements Room Temperature
Product Type Biochemicals
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.