missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INF 39
Description
INF 39 is an irreversible NLRP3 inhibitor (exhibits 52% inhibition of NLRP3 ATPase at 100 μM). Inhibits IL-1β release, caspase activation and pyroptosis in THP-1 cells. Reduces inflammation in a rat colitis model. Exhibits low cytotoxicity.
Specifications
Specifications
| Chemical Name or Material | Ethyl 2-(2-chlorobenzyl)acrylate |
| CAS | 866028-26-4 |
| Assay Percent Range | >98% |
| Quantity | 10 mg |
| Product Line | Tocris |
| Molecular Formula | C12H13ClO2 |
| Formula Weight | Observed MW: 224.68 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?