missing translation for 'onlineSavingsMsg'
Learn More

INF39, MedChemExpress

Product Code. 16494706
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16494706 100 mg 100mg
16460017 50 mg 50mg
16452796 25 mg 25mg
16433628 10 mg 10mg
16406357 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16494706 Supplier Medchem Express Supplier No. HY101868
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress INF39 is an irreversible and noncytotoxic NLRP3 inhibitor.

INF39 is an irreversible and noncytotoxic NLRP3 inhibitor.

Specifications

Chemical Name or Material INF39
CAS 866028-26-4
Color Colorless
Purity Grade Notes Research
Molecular Formula C12H13ClO2
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (445.08 mM; Need ultrasonic)
SMILES O=C(OCC)C(CC1=CC=CC=C1Cl)=C
Molecular Weight (g/mol) 224.68
Formula Weight 224.68
Percent Purity 99.23%
Grade Research
Physical Form Liquid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.