missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences Influenza M peptide GILGFVFTL (HLA-A*0201)

Single peptide synthesized as presented by the MHC class I allele

Brand:  IBA Lifesciences 6-7003-901

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15342176

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Influenza M peptide GILGFVFTL (HLA-A*0201) is a single peptide for stimulation of T cells.
  • The peptide from Influenza M is synthesized as it is presented by the MHC class I HLA-A*0201 allele and can be used for targeted in vitro generation and expansion of antigen-specific CD8+ T cells.
  • The peptides were purified by HPLC using a water and acetonitrile gradient and are endotoxin-free.
  • Specifications

    Specifications

    Recommended: -25°C to -15°C
    1 mg
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers