missing translation for 'onlineSavingsMsg'
Learn More

Ingenol-3,4,5,20-diacetonide, MedChemExpress

Product Code. 16472559
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16472559 1 mg 1mg
16471558 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16472559 Supplier Medchem Express Supplier No. HYN0871

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

MedChemExpress Ingenol-3,4,5,20-diacetonide is a natural compound.

Ingenol-3,4,5,20-diacetonide is a natural compound.

Specifications

Chemical Name or Material Ingenol-3,4,5,20-diacetonide
CAS 77573-44-5
Color White
Purity Grade Notes Research
Molecular Formula C26H36O5
Quantity 1 mg
Synonym Ingenol 3,4:5,20-bisacetonide
SMILES O=C1[C@@]2(C=C(C)C3OC(C)(C)O4)C34C(OC(C)(C)OC5)C5=C[C@@H]1[C@@H](C6(C)C)[C@@H]6CC2C
Molecular Weight (g/mol) 428.56
Formula Weight 428.56
Percent Purity 98.19%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.