missing translation for 'onlineSavingsMsg'
Learn More

Iniparib, Thermo Scientific™

Product Code. 17545575 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Product Code. Quantity unitSize
17545575 100 mg 100mg
17555575 500 mg 500mg
2 options
This item is not returnable. View return policy

Product Code. 17545575

Brand: Thermo Scientific Acros 469161000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 160003-66-7
Molecular Formula C7 H5 I N2 O3

Specifications

Quantity 100 mg
Formula Weight 298.30
MDL Number MFCD11110639
Physical Form Powder
Color White to Beige
Purity ≥97.5%
InChI Key MDOJTZQKHMAPBK-UHFFFAOYSA-N
SMILES NC(=O)C1=CC(=C(I)C=C1)[N+]([O-])=O
IUPAC Name 4-iodo-3-nitrobenzamide
Molecular Weight (g/mol) 292.03
Chemical Name or Material Iniparib
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.