missing translation for 'onlineSavingsMsg'
Learn More

Miele Professional™ Injector nozzle

Product Code. 10518573 Shop All Miele Products
Change view
Click to view available options
Diameter (Metric):
6 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
10518573 6 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10518573 Supplier Miele Professional™ Supplier No. 3809510
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Injector nozzles E 352 for the optimum cleaning of the wash items

  • Suitable for use in a mobile injector unit
  • Can be combined with clip E 354
  • Ø 6 x 220 mm, screw connection
TRUSTED_SUSTAINABILITY

Specifications

Material Stainless Steel
Product Type Injector Nozzle
Color Stainless Steel
For Use With (Equipment) Labwasher
Diameter (Metric) 6 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.