missing translation for 'onlineSavingsMsg'
Learn More

Miele Professional™ Injector Nozzle

Product Code. 10486402 Shop All Miele Products
Change view
Click to view available options
Model:
ID 110
ID 140
ID 160
ID 180
ID 240
ID 90
Dimensions (Dia. x H):
2.5 dia. x 110mm L
2.5 dia. x 90mm L
4 dia. x 140mm L
4 dia. x 160mm L
4 dia. x 180mm L
6 dia. x 240 mm L
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Dimensions (Dia. x H) unitSize
10486402 ID 90 2.5 dia. x 90mm L Each
10008441 ID 110 2.5 dia. x 110mm L Each
10764523 ID 140 4 dia. x 140mm L Each
10712993 ID 160 4 dia. x 160mm L Each
10231572 ID 180 4 dia. x 180mm L Each
10640384 ID 240 6 dia. x 240 mm L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 10486402 Supplier Miele Professional™ Supplier No. 3810320
Estimated Shipment: 16-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Injector nozzles with plastic supports

  • Suitable for standard mobile injector units and injector baskets
  • Suitable for measuring flasks and test tubes, for example
TRUSTED_SUSTAINABILITY

Specifications

Material Stainless Steel
Product Type Injector Nozzle
Color Stainless Steel
Model ID 90
For Use With (Equipment) Washer-disinfectors, laboratory
Includes Plastic support
Dimensions (Dia. x H) 2.5 dia. x 90mm L
Quantity 1 Each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.