missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ Ink Ribbon Cartridges

Product Code. 12693506
Change view
Click to view available options
Color:
Black
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
12693506 Black Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12693506 Supplier Mettler Toledo™ Supplier No. 44000250
  • ONLINE EXCLUSIVE PRICE
    181.00 €
    150.00 € / Pack of 10
    Save 31.00 €
    17% Off
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

Generate immediate documentation of results. Mettler Toledo™ Ink Ribbon Cartridges are used with a variety of dot matrix printers that can be attached to a range of balances, scales and meters. Quality ink on paper rolls provides an instant, clear print-out for reliable record-keeping.

Many Mettler Toledo™ balances, scales and meters can be attached directly to small dot-matrix printers for instant documentation of results using a minimum of benchtop space. These printers use roller paper and small ink cartridges.

Set of 10 replacement ink cartridges for the following dot matrix printers:

  • LCP-43
  • RS-P42
  • Sprinter
  • LCP-45
  • SQC-14
TRUSTED_SUSTAINABILITY

Specifications

Color Black
For Use With (Equipment) Mettler Toledo LCP-43 Printer, RS-P42 Printer, Sprinter, LCP-45 Printer, SQC-14 Printer
Print Technology Dot Matrix
Format Cartridge, Ribbon
For Use With (Application) Instant documentation of results using Mettler Toledo™ meters, scales and balances
Quantity 10 Cartidges
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.