Learn More
Mettler Toledo™ InLab™ DES™ Cable
GREENER_CHOICE
Product Code. 31454080
Change view
Click to view available options
Length (Metric) Cable:
1.2 m
3 m
5 m
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Cable
31454080 1.2 m
31454046 3 m
31454050 5 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 31454080 Supplier Mettler Toledo™ Supplier No. 31052331
In Stock
Add to basket
Add to basket
Digital cable designed for secure connection with DES sensors, ensuring stable signal transmission and reliable measurement results.
  • One cable for all digital parameters
  • DES™ - Diagnosable Efficient Sustainable:
    • Increased efficiency with sensor diagnostics
    • Faster problem solving, less downtime, maximized sensor lifespan, and support of preventive maintenance.
  • Stay ahead: The sensor's diagnostic capability monitors sensor condition, supports preventive maintenance, maximizes the sensor's lifespan, and prevents premature discard
  • Sustainable investment: DES™ digital sensors are a smart choice. Retaining the measurement electronics and only replacing the sensor shaft at the end of its life, or the detachable cable that can be used for all sensors, helps reduce material waste and thus, the ecological footprint
  • Boost productivity and efficiency: With diagnosed errors, the user guidance with troubleshooting recommendations enables faster problem-solving, reducing downtime. It also minimizes handling errors, thereby cutting down on the need for specialized training
TRUSTED_SUSTAINABILITY
Specifications
Type Cable
Length (Metric) Cable 1.2 m