missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ InLab™ Max Pro-ISM pH Electrode

Product Code. 15519611 Shop All Mettler Toledo Products
Change view
Click to view available options
Connector Types:
Multi-Pin
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connector Types unitSize
15519611 Multi-Pin Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15519611 Supplier METTLER TOLEDO™ Supplier No. 30248830
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

An all-purpose pH electrode featuring an immovable glass sleeve to help prevent pollutants from entering and clogging the junction.

  • For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
  • Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • High-ion strength
  • High temperature samples
  • Wastewater
  • TRIS buffers
  • Pharmaceutical & Biological samples
  • Organic solvents
  • Fruit juice
  • Water
  • General lab

Accessories & Care

  • Immovable glass sleeve junction
  • Refillable 3M KCl (51350072) - Keep probe filled with electrolyte for accurate results
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)

Recommended Storage

InLab storage solution

TRUSTED_SUSTAINABILITY

Specifications

Connector Types Multi-Pin
Junction Immovable Glass Sleeve
Material Glass Shaft
pH Range 0 to 14
Type Combination Electrode; Integrated Temperature Probe; Intelligent Sensor Management (ISM) Technology
For Use With (Application) General laboratory applications
For Use With (Equipment) Multi-Pin to BNC/RCA (30281896)
Internal Reference ARGENTHAL™with Ag+-trap
Membrane HA Glass
Model InLab™ Max Pro-ISM
Storage Requirements InLab storage solution
Temperature (Metric) Range 0°C to 100°C
Length (Metric) Stem 120 mm
Diameter (Metric) Body 12 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.