missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- For use with METTLER TOLEDO Seven2Go and SevenGoDuo portable pH meters.
- Capture electrode calibration and results with EasyDirect software.
Featured Applications
- Low-ion strength (pure water)
- Water
- Field/Environmental sampling
Accessories & Care
- Porous PTFE junction
- Refillable 3M KCl (51350072): Keep probe filled with electrolyte for accurate results
- Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
- Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
Recommended Storage
InLab storage solution
Specifications
Specifications
| Connector Types | BNC, RCA IP67 |
| Junction | Porous PTFE |
| Material | Polysulfone Shaft |
| pH Range | 1 to 11 |
| Type | Combination Electrode; Integrated Temperature Probe (NTC 30 kΩ) |
| For Use With (Application) | Measuring samples at low temperatures or with low ion concentrations |
| For Use With (Equipment) | Fixed cable to sensor |
| Internal Reference | ARGENTHAL™ with Ag+-trap |
| Membrane | LoT Glass |
| Model | InLab™ Water Go |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?