Learn More
Cytiva Inlet Tubing Kit
Product Code. 11613224 Shop All Cytiva Products
Change view
Click to view available options
Includes:
5 x Tubing (5/16 male / 1/8 in.) I.D. 1.6 mm, O.D. 3.15 mm (1/8 in.), length 1.0 m, Unions, Ferrules
Unit Size:
Each
Product Code. Includes Unit Size
11613224 5 x Tubing (5/16 male / 1/8 in.) I.D. 1.6 mm, O.D. 3.15 mm (1/8 in.), length 1.0 m, Unions, Ferrules Each
1 options
Product Code. 11613224 Supplier Cytiva Supplier No. 28957215
Estimated Shipment: 20-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Description Inlet Tubing Kit
Material Fluoroethenepropene (FEP)
Includes 5 x Tubing (5/16 male / 1/8 in.) I.D. 1.6 mm, O.D. 3.15 mm (1/8 in.), length 1.0 m, Unions, Ferrules