missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Inner Lens Cover
Description
- Replacement inner lens cover, size 90 x 110 mm
- For use with 3M™ Helmets, HT-Series
- Genuine 3M™ Accessory
Specifications
Specifications
| Certifications/Compliance | CE Approved |
| Dimensions (L x W) | 90 x 100 mm |
| For Use With (Equipment) | Helmet HT-Series |
| Product Type | Inner Lens Cover |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?