missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ InnoviGene™ Suite Software

Product Code. 31453978 Shop All Applied Biosystems Products
Change view
Click to view available options
Product Type:
Fragment Analysis Software
Fragment Analysis Software Renewal
Sanger Sequencing & Fragment Analysis Software
Sanger Sequencing & Fragment Analysis Software Renewal
Sequencing Software
License:
1 Year
3 Years
5 Years
Unit Size:
Each
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type License Unit Size
31453978 Fragment Analysis Software Renewal 5 Years -
31453971 Fragment Analysis Software 1 Year -
31453965 Fragment Analysis Software 3 Years -
31453969 Fragment Analysis Software 5 Years -
31453958 Fragment Analysis Software Renewal 1 Year -
31453960 Fragment Analysis Software Renewal 3 Years -
31453966 Sanger Sequencing & Fragment Analysis Software 1 Year -
31453975 Sanger Sequencing & Fragment Analysis Software 3 Years -
31453972 Sanger Sequencing & Fragment Analysis Software 5 Years -
31453974 Sanger Sequencing & Fragment Analysis Software Renewal 1 Year -
31453961 Sanger Sequencing & Fragment Analysis Software Renewal 3 Years -
31453973 Sanger Sequencing & Fragment Analysis Software Renewal 5 Years -
18892544 Sequencing Software 1 Year Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
This item is not returnable. View return policy
Product Code. 31453978 Supplier Applied Biosystems™ Supplier No. A40004739
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

InnoviGene Suite software enables you to analyze and manage capillary electrophoresis sequencing & fragment analysis data generated by Applied Biosystems genetic analyzers.

Applied Biosystems InnoviGene Suite software enables you to analyze and manage capillary electrophoresis sequencing and fragment analysis data generated by Applied Biosystems genetic analyzers. The software provides a streamlined workflow to guide you through uploading and analyzing the data and generating reports.

InnoviGene Suite includes SAE administrator console software with security, audit, and electronic signature (SAE) functions that support compliance with guidelines on electronic records security and prevents unauthorized access to the instrument.

The software is delivered via a download link sent to you after purchase.

Applications within InnoviGene Suite

InnoviGene Suite allows you to create projects for use with other applications that are integrated into the software. These applications enable you to complete an end-to-end workflow using projects that contain related files. The applications available depend on the type of InnoviGene Suite purchased ('sequencing' or 'fragment analysis').

Sequence Quality Check application (Innovigene Suite for sequencing)—enables you to perform the following activities:

  • View the quality information for trace files
  • Change quality settings and reanalyze trace files
  • Perform comparative analysis across multiple trace files

Sequence Identity application (InnoviGene Suite for sequencing)—enables you to view the assembly of multiple files compared to a reference and view basecall mismatches. Specifically, you can perform the following activities with the application:

  • Basecalling, quality value assignment, and mixed base identification
  • Trimming low-quality ends from each sequence, if the setting is active
  • Identifying poor-quality samples
  • Assembling the samples against the reference sequence and generating a specimen consensus sequence
  • Reviewing the basecalling quality values and consensus quality values
  • Identifying mismatches by aligning and comparing specimen sequences to the reference sequence. A mismatch is a position in the specimen that does not match with the reference sequence. Mismatches include base changes, insertions, and deletions.
  • Assigning specimen status based on alignment with the reference sequence and coverage criteria
  • Editing basecalls in samples or the consensus
  • Generating reports and results for export in CSV format

STR Typing application (InnoviGene Suite for fragment analysis)—the STR Typing application is genotyping software that uses short tandem repeat (STR) data. It enables you to perform the following activities:

  • Analyze a mixture of DNA fragments, separated by size
  • Provide a profile of the separation
  • Calculate the sizes of the fragments
  • Determine the alleles present in fragment analysis sample (FSA) files
TRUSTED_SUSTAINABILITY

Specifications

Sequencing Type Fragment Analysis
For Use With (Application) Fragment Analysis
Quantity 5 Users
Product Line InnoviGene
Format Software
For Use With (Equipment) Applied Biosystems™ Genetic Analyzers
License 5 Years
Product Type Fragment Analysis Software Renewal

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.