missing translation for 'onlineSavingsMsg'
Learn More

Heathrow Scientific™ Inoculating Needle/Loops

Product Code. 16310786 Shop All Thermo Scientific CO₂ Incubators Products
Change view
Click to view available options
Volume:
1 μL
10 μL
Material:
ABS
Polypropylene
Unit Size:
Pack of 1000
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume Material Unit Size
16310786 10 μL ABS Pack of 1000
16380776 1 μL Polypropylene Pack of 1000
16390776 1 μL ABS Pack of 1000
16300786 10 μL Polypropylene Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16310786 Supplier Heathrow Scientific™ Supplier No. HS81121D
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Provide the choice to spread or pick colonies and cultures without the risk of contamination with Inoculating Needle/Loops.

  • Use loop end to uniformly streak a gel surface
  • Use needle end for removal of colonies
  • Lot-to-lot traceability
  • Sterile
  • Disposable

Specifications

Disposable Single-use
Volume 10 μL
Length (English) 7.9 in.
Length (Metric) 20 cm
Material ABS
Sterility Sterile
Size Standard
Color Blue
Product Type Inoculating Loop and Needle, Rigid
Autoclavable Not Autoclavable
For Use With (Application) Spreading, Picking, and Transferring Colonies or Cultures
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.