missing translation for 'onlineSavingsMsg'
Learn More

WTW™ inoLab™ 7310 Benchtop pH Meter with Precision Electrode
GREENER_CHOICE

Product Code. 12661685
Change view
Click to view available options
Display Type:
LCD, Graphic with Backlight
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Display Type unitSize
12661685 LCD, Graphic with Backlight Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12661685 Supplier WTW™ Supplier No. 1AA314
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

A precise pH meter with documentation function to use in quality assurance, the pharmaceutical industry and everywhere GLP-supported documentation is required.

  • Repeatable measuring results by automatic recognition of stable values
  • Monitoring of the optimum measuring range vai CMC function
  • Secure and convenient operation via text menus in a bright, backlit graphic display
  • All measured values are transmitted with the current date, time, ID number and sensor serial number
  • Provided with a combined precision pH electrode
TRUSTED_SUSTAINABILITY

Specifications

Includes Combined pH electrode. Meter with universal power supply, stand and operation manual. Combined pH electrode SenTix™ 81, Buffer 4, 7 and 10.01, 3mol/L KCl, software and USB cable, WTW technical Buffers, DIN/NIST Buffers plus 20 additional Buffer sets
Accuracy (Temperature) ±0.1K
Accuracy (mV) ±0.3, ±1
Accuracy (pH) ±0.005, ±0.01
Data Management Details Up to 10 records calibration memory, 500 manual datasets/5000 automatic memory
For Use With (Application) For measurements/documentation according GLP/AQA, AC and battery operation
Range (pH) -2 to 20
Resolution (pH) -2 to 20 ±0.01, -2 to 19.999 ±0.005
Range (mV) ±1200, ±0.3
Resolution (mV) ±(1200 ±1)mV
Calibration Points 1 to 3
Display Type LCD, Graphic with Backlight
Certifications/Compliance CE, cETLUS
Dimensions (L x W x H) 240 x 190 x 80 mm
Weight (Metric) 0.8 kg
Warranty 3 Years
Power Supply Four 1.5V AA batteries or four 1.2V NiMH
Relative Humidity Range Yearly mean: <75%, 30 days/year: 95%, Other days: 85%
Temperature Compensation Automatic/Manual
Temperature (Metric) Operating -5°C to +105°C
Temperature Sensor DIN or BNC/4mm banana
Type inoLab™ Wastewater 7310 set 4 pH meter
Battery Life Up to 1000 hr. without backlight, 150 hr. with backlight
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.