missing translation for 'onlineSavingsMsg'
Learn More

Inolitazone dihydrochloride, MedChemExpress

Product Code. 16488767
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488767 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16488767 Supplier Medchem Express Supplier No. HY14792B
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Inolitazone dihydrochloride (Efatutazone dihydrochloride) is a novel high-affinity PPARγ agonist that is dependent upon PPARγ for its biological activity with IC50 of 0.8 nM for growth inhibition.

Inolitazone dihydrochloride (Efatutazone dihydrochloride) is a novel high-affinity PPARγ agonist that is dependent upon PPARγ for its biological activity with IC50 of 0.8 nM for growth inhibition.

Specifications

Chemical Name or Material Inolitazone dihydrochloride
CAS 223132-38-5
Color Pale Purple
Purity Grade Notes Research
Molecular Formula C27H28Cl2N4O4S
Quantity 5 mg
Synonym Efatutazone dihydrochlorideCS-7017 dihydrochlorideRS5444 dihydrochloride
Solubility Information DMSO : 25 mg/mL (43.44 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(N1)SC(CC2=CC=C(OCC3=NC4=CC=C(OC5=CC(C)=C(N)C(C)=C5)C=C4N3C)C=C2)C1=O.Cl.Cl
Molecular Weight (g/mol) 575.51
Formula Weight 575.51
Percent Purity 98.36%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.