missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Inosine, 98+%

Product Code. 11491768
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11491768 100 g 100g
11471768 5 g 5g
11481768 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11491768 Supplier Thermo Scientific Chemicals Supplier No. A14459.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 58-63-9 | C10H12N4O5 | 268.23 g/mol

Suppresses the increase of glucose and insulin in the blood

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-63-9
Molecular Formula C10H12N4O5
Molecular Weight (g/mol) 268.23
MDL Number MFCD00066770
InChI Key UGQMRVRMYYASKQ-YPLCUDRINA-N
Synonym inosine, hypoxanthosine, ribonosine, atorel, oxiamin, trophicardyl, selfer, pantholic-l, panholic-l, hypoxanthine riboside
PubChem CID 6021
ChEBI CHEBI:17596
IUPAC Name 9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
SMILES OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=NC2=C1N=CNC2=O

Specifications

Melting Point 212°C to 215°C (decomposition)
Quantity 100 g
Beilstein 624889
Merck Index 14,4975
Optical Rotation −52° (c=1 in Water)
Formula Weight 268.23
Percent Purity ≥98%
Chemical Name or Material Inosine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.