missing translation for 'onlineSavingsMsg'
Learn More

Inosine, 99%

Product Code. 10152490 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
250 g
Packaging:
Glass bottle
Unit Size:
100g
10g
250g
25g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10152490 10 g Glass bottle 10g
10396560 25 g Glass bottle 25g
10794431 100 g Glass bottle 100g
10553884 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10152490 Supplier Thermo Scientific Acros Supplier No. 122250100
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 58-63-9 | C10H12N4O5 | 268.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-63-9
Molecular Formula C10H12N4O5
Molecular Weight (g/mol) 268.23
MDL Number MFCD00066770
InChI Key UGQMRVRMYYASKQ-YPLCUDRINA-N
Synonym inosine, hypoxanthosine, ribonosine, atorel, oxiamin, trophicardyl, selfer, pantholic-l, panholic-l, hypoxanthine riboside
PubChem CID 6021
ChEBI CHEBI:17596
IUPAC Name 9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
SMILES OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=NC2=C1N=CNC2=O

Specifications

Absorbance 1.65 to 1.77 (A250/A260), 0.22 to 0.26 (A280/A260)
Melting Point 212°C to 213°C
Color White
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 10 g
Beilstein 31, 25
Merck Index 15, 5018
Specific Rotation Condition − 49.20 (18.00°C c=1,H2O)
Specific Rotation -49.2
Solubility Information Solubility in water: 2.1g/100 mL (20°C). Other solubilities: soluble in ethanol
Spectrophotometry 98 to 102% (lambda max. 248.5nm,pH 7) Molar absorptivity 12200+/-2%,on dry substance
Transmittance 98% min. (c=1, H2O, 430nm, 1cm cell)
Formula Weight 268.22
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Inosine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.