missing translation for 'onlineSavingsMsg'
Learn More

Inosine, MedChemExpress

Product Code. 16488858
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
100 g
Unit Size:
100g
10g
1g
25g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16488858 10 g 10g
16472489 1 g 1g
16468976 100 g 100g
16453836 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16488858 Supplier Medchem Express Supplier No. HYN0092
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Inosine is an endogenous purine nucleoside produced by catabolism of adenosine. Inosine has anti-inflammatory, antinociceptive, immunomodulatory and neuroprotective effects. Inosine is an agonist for adenosine A1 (A1R) and A2A (A2AR) receptors.

Inosine is an endogenous purine nucleoside produced by catabolism of adenosine. Inosine has anti-inflammatory, antinociceptive, immunomodulatory and neuroprotective effects. Inosine is an agonist for adenosine A1 (A1R) and A2A (A2AR) receptors.

Specifications

Chemical Name or Material Inosine
CAS 58-63-9
Color White
Purity Grade Notes Research
Molecular Formula C10H12N4O5
Quantity 10 g
Solubility Information DMSO : 25 mg/mL (93.20 mM; Need ultrasonic) ∣H2O : 10 mg/mL (37.28 mM; Need ultrasonic)
SMILES OC[C@@H]1[C@H]([C@H]([C@H](N2C=NC3=C2N=CN=C3O)O1)O)O
Molecular Weight (g/mol) 268.23
Formula Weight 268.23
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.