missing translation for 'onlineSavingsMsg'
Learn More

(-)-Inosine, TRC

Product Code. 30012781
Change view
Click to view available options
Quantity:
1 g
50 g
1 kg
Unit Size:
1g
1kg
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012781 50 g 50g
30009142 1 kg 1kg
17909274 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012781 Supplier Toronto Research Chemicals Supplier No. TRCI66100050G
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-63-9
Molecular Formula C10 H12 N4 O5
Molecular Weight (g/mol) 268.23
Synonym 9-β-D-Ribofuranosyl-1,9-dihydro-6H-purin-6-one, Adenosine Imp. G (EP), Inosine, Didanosine Imp. B (Ph. Int.)
IUPAC Name 9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1H-purin-6-one
SMILES OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)n2cnc3C(=O)NC=Nc23

Specifications

Quantity 50 g
InChI Formula InChI=1S/C10H12N4O5/c15-1-4-6(16)7(17)10(19-4)14-3-13-5-8(14)11-2-12-9(5)18/h2-4,6-7,10,15-17H,1H2,(H,11,12,18)/t4-,6-,7-,10-/m1/s1
Formula Weight 268.0808
Percent Purity >95%
Chemical Name or Material Inosine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.