missing translation for 'onlineSavingsMsg'
Learn More

Miele Professional™ Insert

Product Code. 10452773 Shop All Miele Products
Change view
Click to view available options
Depth (Metric):
429 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Depth (Metric) Unit Size
10452773 429 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10452773 Supplier Miele Professional™ Supplier No. 12666290
Estimated Shipment: 05-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Insert A 11/1 for the gentle cleaning of delicate items by interrupting the spray jet

  • With perforations 7 x 7 mm
  • For use in upper and lower basket
TRUSTED_SUSTAINABILITY

Specifications

Material Stainless Steel
Product Type Insert
Color White
For Use With (Equipment) Washer-disinfectors, laboratory
Depth (Metric) 429 mm
Width (Metric) 429 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.