missing translation for 'onlineSavingsMsg'
Learn More

Miele Professional™ Insert

Product Code. 12007076 Shop All Miele Products
Change view
Click to view available options
Dimensions (L x W x H):
445 x 180 x 110 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) Unit Size
12007076 445 x 180 x 110 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12007076 Supplier Miele Professional™ Supplier No. 3203500
Estimated Shipment: 30-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Insert E 379 for holding different instruments

  • 0.8 mm wire mesh, 1.7 mm mesh width, 5 mm frames
  • equipped with 2 handles
  • For use in upper and lower basket
TRUSTED_SUSTAINABILITY

Specifications

Material Stainless Steel
Color Stainless Steel
For Use With (Equipment) Washer-disinfectors, medical
Dimensions (L x W x H) 445 x 180 x 110 mm
Height (Metric) 110 mm
Length (Metric) 445 mm
Width (Metric) 180 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.