missing translation for 'onlineSavingsMsg'
Learn More

Beckman Coulter™ Insert for IgG Purity/Heterogeneity Assay Kit

Product Code. 10680376 Shop All Beckman Coulter Life Sciences Products
Encompass
Change view
Click to view available options
Kit Size:
200 Runs
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kit Size Unit Size
10680376 200 Runs Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10680376 Supplier Beckman Coulter™ Supplier No. A10663
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with capillary cartridge systems with a photodiode array (PDA) detector (e.g. PA 800 systems). Beckman Coulter™ Insert for IgG Purity/Heterogeneity Assay Kit contains the capillaries and all the reagents needed to perform 200 runs.

The Beckman Coulter IgG Purity/Heterogeneity Assay Kit is used to resolve both reduced and nonreduced immunoglobulins by size, and to quantify the heterogeneity and impurities that may exist in a given IgG preparation.

Kit components:

  • Capillary, 50μm I.D. bare-fused silica: 2 capillaries
  • SDS-MW Sample Buffer, 100mM Tris-HCl, pH 9.0, 1% SDS: 50mL
  • SDS-MW Gel Buffer, pH 8, 0.2% SDS: 140mL 4-pack
  • IgG Control Standard, 1mg/mL: 1mL 3-pack
  • Internal Standard, 10kDa protein, 5mg/mL: 0.4mL
  • Acidic Wash Solution, 0.1M HCl: 100mL
  • Basic Wash Solution, 0.1M NaOH: 100mL

Specifications

Includes 2 x Capillary, SDS-MW Sample Buffer, SDS-MW Gel Buffer, Internal Standard (10 kDa), IgG Control Standard, Acid Wash Solution, Basic Wash Solution
Kit Size 200 Runs
For Use With (Application) Resolve immunoglobulins by size, and quantify the heterogeneity and impurities
Storage Requirements Room temperature (15-30°C), except for IgG Control Standard (-35 to -15°C), Internal Standard (2-8°C)
Format Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.