Learn More
Cytiva Insert Release
Product Code. 17154721
Change view
Click to view available options
Volume (Metric):
20 to 60 mL (Sample)
Unit Size:
Pack of 24
Product Code. Volume (Metric) unitSize
17154721 20 to 60 mL (Sample) Pack of 24
1 options
Product Code. 17154721
Brand: Cytiva FD003X65
Only null left
Add to basket
Add to basket
Insert release for use with protein purification and sterile filtration workflows.
  • Utilizes proprietary high-performance multi-layer filter membranes
  • Plates offer time savings strong performance claims and streamlined workflow improvements
  • Each plate comes with a V-shaped bottom collection plate and lid
TRUSTED_SUSTAINABILITY
Specifications
Description EFA: 15.2 cm2
For Use With (Equipment) Jumbosep Centrifugal Device
Material (Membrane) PES
Sterility Non-sterile
Housing Material Polypropylene
Quantity 24/Pk.
Volume (Metric) 20 to 60 mL (Sample)