Learn More
Abeba™ Insoles 3551
Product Code. 15600118
Change view
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15600118 35 1 pair
15610118 36 1 pair
15620118 37 1 pair
15630118 38 1 pair
15640118 39 1 pair
15650118 40 1 pair
15660118 41 1 pair
15670118 42 1 pair
15680118 43 1 pair
15690118 44 1 pair
15600128 45 1 pair
15610128 46 1 pair
15620128 47 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 15600118 Supplier Abeba™ Supplier No. 3551/35
Only null left
Add to basket
Add to basket
Replaceable insole for shoes with air cushion sole is ESD dissipative and antistatic.
  • Gray
  • Antistatic
  • ESD dissipative according to DIN EN 61340
  • Replaceable insole for shoes with air cushion sole
Specifications
Color Gray
Size 35
Style 3551