Learn More
Abeba™ Insoles 3566
Product Code. 15610208
Change view
Click to view available options
Size:
40
41
42
43
44
45
46
Unit Size:
1 pair
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15610208 40 1 pair
15620208 41 1 pair
15630208 42 1 pair
15640208 43 1 pair
15650208 44 1 pair
15660208 45 1 pair
15670208 46 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 15610208 Supplier Abeba™ Supplier No. 3566/40
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
Massage knobs stimulate circulation. ESD dissipative and antistatic.
  • Microfiber
  • ESD dissipative according to DIN EN 61340
  • Antistatic
  • Stimulating circulation by massage knobs, comfortable orthopaedic footbed
  • Reflexor massage active-footbed
Specifications
Material Microfiber
Size 40
Style 3566